Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145826.1 | NP_001139298.1 | 6 - 7 | 723 | 68 | |
NM_153406.3 | NP_700455.3 | 3 - 4 | 494 | 68 | ||
GenBank mRNA | AB093236.1 | - | 6 - 7 | 723 | 68 | |
AK035500.1 | - | 3 - 4 | 496 | 68 | ||
AK052716.1 | - | 3 - 4 | 495 | 68 | ||
AY884297.1 | - | 3 - 4 | 495 | 68 | ||
BC072595.1 | - | 2 - 3 | 333 | 6408832 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |