Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081688.2 | NP_001075157.2 | 16 - 17 | 3002 | 91 | |
GenBank mRNA | BC158102.1 | - | 16 - 17 | 3098 | 9007387 | 91 |
BC172169.1 | - | 16 - 17 | 3098 | 9054839 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |