Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177769.4 | NP_808437.2 | 5 - 6 | 652 | 110 | |
GenBank mRNA | AK029207.1 | - | 5 - 6 | 533 | 109 | |
AK132239.1 | - | 5 - 6 | 653 | 110 | ||
BC049160.1 | - | 5 - 6 | 515 | 110 | ||
BC120566.1 | - | 4 - 5 | 368 | 110 | ||
BC144961.1 | - | 4 - 5 | 368 | 110 | ||
BC144963.1 | - | 4 - 5 | 330 | 9053511 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |