Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168624.1 | NP_001162095.1 | 1 - 2 | 136 | 82 | |
GenBank mRNA | AK029946.1 | - | 1 - 2 | 137 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |