Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190911.1 | NP_001177840.1 | 3 - 4 | 649 | 60 | |
NM_001190912.1 | NP_001177841.1 | 2 - 3 | 571 | 60 | ||
NM_001190913.1 | NP_001177842.1 | 3 - 4 | 649 | 60 | ||
NM_001190914.1 | NP_001177843.1 | 4 - 5 | 750 | 60 | ||
NM_026324.3 | NP_080600.1 | 3 - 4 | 649 | 60 | ||
GenBank mRNA | AK034142.1 | - | 3 - 4 | 586 | 60 | |
AK038385.1 | - | 3 - 4 | 400 | 60 | ||
AK045373.1 | - | 3 - 4 | 554 | 60 | ||
AK079332.1 | - | 4 - 5 | 687 | 60 | ||
AK122566.1 | - | 2 - 3 | 163 | 60 | ||
AY169782.1 | - | 3 - 4 | 649 | 60 | ||
BC063072.1 | - | 2 - 3 | 366 | 5695003 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |