Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025612.2 | NP_001020783.1 | 4 - 5 | 411 | 69 | |
GenBank mRNA | AK032028.1 | - | 4 - 5 | 400 | 69 | |
BC138950.1 | - | 4 - 5 | 398 | 8861972 | 69 | |
BC138951.1 | - | 4 - 5 | 398 | 8861973 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |