Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144922.2 | NP_659171.1 | 10 - 11 | 1605 | 85 | |
GenBank mRNA | AK087545.1 | - | 10 - 11 | 1575 | 85 | |
AK155604.1 | - | 10 - 11 | 1616 | 85 | ||
AK182216.1 | - | 1 - 2 | 67 | 85 | ||
BC021506.1 | - | 10 - 11 | 1629 | 5347099 | 85 | |
BC027844.1 | - | 7 - 8 | 898 | 85 | ||
BC078642.1 | - | 10 - 11 | 1529 | 6329630 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |