Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009022.4 | NP_033048.2 | 10 - 11 | 1307 | 122 | |
GenBank mRNA | AK078553.1 | - | 10 - 11 | 1248 | 122 | |
BC075704.1 | - | 10 - 11 | 1274 | 30471325 | 122 | |
X99273.1 | - | 10 - 11 | 1252 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |