Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900007598.mm.1 |
NM_001163725.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007598.mm.1 |
NM_001163725.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007598.mm.1 |
NM_026314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007598.mm.1 |
NM_026314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007598.mm.1 |
TC0900003325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007601.mm.1 |
NM_001163725.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007601.mm.1 |
NM_001163725.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007601.mm.1 |
NM_026314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007601.mm.1 |
NM_026314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007601.mm.1 |
TC0900003325.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900028033.mm.1 |
NM_001163725.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028033.mm.1 |
NM_001163725.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028033.mm.1 |
NM_026314.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028033.mm.1 |
NM_026314.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900028033.mm.1 |
TC0900003325.mm.1 |
MTA TC |