Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177158.5 | NP_796132.2 | 3 - 4 | 279 | 73 | |
GenBank mRNA | AK030993.1 | - | 3 - 4 | 272 | 73 | |
BC044761.1 | - | 3 - 4 | 315 | 73 | ||
BC046530.1 | - | 3 - 4 | 315 | 5329601 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |