Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010706.2 | NP_034836.1 | 6 - 7 | 941 | 69 | |
GenBank mRNA | AF026795.1 | - | 5 - 6 | 491 | 69 | |
AF510729.1 | - | 6 - 7 | 590 | 69 | ||
AK145734.1 | - | 6 - 7 | 761 | 69 | ||
AY044870.1 | - | 6 - 7 | 613 | 69 | ||
BC011236.1 | - | 6 - 7 | 579 | 4205669 | 69 | |
BC021632.1 | - | 6 - 7 | 597 | 4218531 | 69 | |
BC030297.1 | - | 6 - 7 | 754 | 4986324 | 69 | |
BC094008.1 | - | 6 - 7 | 592 | 4986911 | 69 | |
BC141106.1 | - | 6 - 7 | 663 | 9055718 | 69 | |
BC145387.1 | - | 6 - 7 | 663 | 9053945 | 69 | |
EF017938.1 | - | 6 - 7 | 941 | 69 | ||
EF017939.1 | - | 6 - 7 | 941 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |