Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900018310.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018310.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018310.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018307.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018307.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018307.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018309.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018309.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018309.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018304.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018304.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018304.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018301.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018301.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018301.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018303.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018303.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018303.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018305.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018305.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018305.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018308.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018308.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018308.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018312.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018312.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018312.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018306.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018306.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018306.mm.1 |
TC0900002196.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900018311.mm.1 |
NM_145403.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018311.mm.1 |
NM_145403.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900018311.mm.1 |
TC0900002196.mm.1 |
MTA TC |