Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opcml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177906.4 | NP_808574.2 | 6 - 7 | 882 | 74 | |
GenBank mRNA | AK158538.1 | - | 6 - 7 | 1204 | 74 | |
BC076581.1 | - | 6 - 7 | 882 | 30542711 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |