Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaja4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021422.4 | NP_067397.1 | 2 - 3 | 479 | 103 | |
GenBank mRNA | AB032401.1 | - | 3 - 4 | 346 | 103 | |
AK016666.1 | - | 1 - 2 | 76 | 103 | ||
AK046476.1 | - | 2 - 3 | 480 | 103 | ||
AK076175.1 | - | 2 - 3 | 480 | 103 | ||
AK141909.1 | - | 2 - 3 | 481 | 103 | ||
AK197546.1 | - | 2 - 3 | 211 | 103 | ||
BC147484.1 | - | 2 - 3 | 327 | 9056231 | 103 | |
BC147486.1 | - | 2 - 3 | 327 | 9056239 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |