Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020561.2 | NP_065586.3 | 6 - 7 | 1052 | 66 | |
GenBank mRNA | AK151683.1 | - | 6 - 7 | 938 | 66 | |
AK152282.1 | - | 6 - 7 | 953 | 66 | ||
BC139288.1 | - | 6 - 7 | 944 | 8862310 | 66 | |
BC139289.1 | - | 6 - 7 | 944 | 8862311 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |