Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158868.1 | - | 9 - 10 | 3095 | 58 | |
AY243848.2 | - | 7 - 8 | 2475 | 58 | ||
BC057615.1 | - | 9 - 10 | 2704 | 6401651 | 58 | |
BC058433.1 | - | 7 - 8 | 1902 | 58 | ||
BC098311.1 | - | 3237 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |