Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013156.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013156.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013156.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013168.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013168.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013168.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013155.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013155.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013155.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013158.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013158.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013158.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013164.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013164.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013164.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013159.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013159.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013159.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013163.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013163.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013163.mm.1 |
TC0400001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013165.mm.1 |
NM_019732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013165.mm.1 |
NM_019732.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013165.mm.1 |
TC0400001651.mm.1 |
MTA TC |