Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159603.1 | NP_001153075.1 | 18 - 19 | 3109 | 111 | |
NM_001159604.1 | NP_001153076.1 | 18 - 19 | 3115 | 111 | ||
NM_001159605.1 | NP_001153077.1 | 16 - 17 | 2827 | 111 | ||
NM_001159606.1 | NP_001153078.1 | 14 - 15 | 2383 | 111 | ||
NM_030722.2 | NP_109647.2 | 18 - 19 | 3118 | 111 | ||
GenBank mRNA | AF321909.1 | - | 18 - 19 | 3088 | 111 | |
AK122205.1 | - | 18 - 19 | 3098 | 111 | ||
AK141921.1 | - | 18 - 19 | 3115 | 111 | ||
AK154059.1 | - | 17 - 18 | 2707 | 111 | ||
AK161135.1 | - | 16 - 17 | 2821 | 111 | ||
AK162742.1 | - | 17 - 18 | 2805 | 111 | ||
AK163977.1 | - | 18 - 19 | 3106 | 111 | ||
BC048174.1 | - | 18 - 19 | 3068 | 6825203 | 111 | |
BC050747.1 | - | 18 - 19 | 3082 | 6593374 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |