Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172546.2 | NP_766134.1 | 1 - 2 | 608 | 79 | |
GenBank mRNA | AK033015.1 | - | 1 - 2 | 609 | 79 | |
AY151137.1 | - | 1 - 2 | 570 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |