Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177364.3 | NP_796338.2 | 3 - 4 | 311 | 56 | |
GenBank mRNA | AB430861.1 | - | 3 - 4 | 233 | 56 | |
AK042261.1 | - | 3 - 4 | 297 | 56 | ||
AK043324.1 | - | 3 - 4 | 283 | 56 | ||
AK089330.1 | - | 3 - 4 | 283 | 56 | ||
AK090119.1 | - | 3 - 4 | 304 | 56 | ||
AK154898.1 | - | 3 - 4 | 269 | 56 | ||
AK163471.1 | - | 3 - 4 | 283 | 56 | ||
AK169978.1 | - | 3 - 4 | 303 | 56 | ||
AK171384.1 | - | 3 - 4 | 283 | 56 | ||
BC115711.1 | - | 3 - 4 | 280 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |