Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005867.1 | NP_001005867.1 | 10 - 11 | 1513 | 85 | |
NM_028601.3 | NP_082877.2 | 10 - 11 | 1609 | 85 | ||
GenBank mRNA | BC024081.1 | - | 9 - 10 | 1359 | 85 | |
BC040745.1 | - | 1 - 2 | 68 | 85 | ||
BC060652.1 | - | 10 - 11 | 1451 | 30354393 | 85 | |
BC063069.1 | - | 2 - 3 | 214 | 85 | ||
CA318375.1 | - | 5 - 6 | 688 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |