Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271160 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271163 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271158 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271161 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271162 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271159 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2170681 |
AY550907.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2170681 |
BC068181.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2170681 |
AK077717.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2170681 |
AK077920.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2170681 |
AK078404.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
652104 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
810422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
400413 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
451651 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489736 |
XM_894554.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489736 |
XM_906003.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
39048 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
439343 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
822433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
900831 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
592567 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
62602 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
37214 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P110084 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P159286 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2170681 |
AY550907.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2170681 |
BC068181.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2170681 |
AK077717.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2170681 |
AK077920.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2170681 |
AK078404.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271159 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271158 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271156 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271161 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271163 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271155 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271154 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23271152 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P110084 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P159286 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00978661 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05388209 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P155142 |
AK082800.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P392544 |
AY550907.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P392544 |
BC068181.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P392544 |
AK077717.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P392544 |
AK077920.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P392544 |
AK078404.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P463220 |
AK031571.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P120688 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138667 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P110084 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P159286 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03602278 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03602283 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03602280 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03602289 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03602288 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271159 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271160 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271158 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271161 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271163 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23271162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271003 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271012 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270999 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271043 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271158 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270988 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271033 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23270989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23271075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271130 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271156 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271088 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271079 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271136 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03915881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271150 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03647985 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03602283 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271129 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271161 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271125 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271085 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271077 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04098169 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271066 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271139 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271067 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271123 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271152 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03953965 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271134 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03574877 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271065 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271131 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03915883 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04098174 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271059 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23271154 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03431126 |
- |
- |