Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009154.mm.1 |
TC0600001187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009154.mm.1 |
NM_001242358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009154.mm.1 |
NM_001242358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
TC0600001187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_001242358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_001242358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_001242360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_001242360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_027296.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009156.mm.1 |
NM_027296.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
TC0600001187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_001242358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_001242358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_001242360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_001242360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_027296.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009155.mm.1 |
NM_027296.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
TC0600001187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_001242358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_001242358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_001242360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_001242360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_027296.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009160.mm.1 |
NM_027296.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
TC0600001187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_001242358.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_001242358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_001242360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_001242360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_027296.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009149.mm.1 |
NM_027296.3 |
Refseq |