Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021134.mm.1 |
TC0600002718.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021134.mm.1 |
NM_001134383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021134.mm.1 |
NM_001134383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021134.mm.1 |
NM_001134384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021134.mm.1 |
NM_001134384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021136.mm.1 |
TC0600002718.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021136.mm.1 |
NM_001134383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021136.mm.1 |
NM_001134383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021136.mm.1 |
NM_001134384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021136.mm.1 |
NM_001134384.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021135.mm.1 |
TC0600002718.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021135.mm.1 |
NM_001134383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021135.mm.1 |
NM_001134383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021135.mm.1 |
NM_001134384.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021135.mm.1 |
NM_001134384.1 |
Refseq |