Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178045.4 | NP_835146.3 | 5 - 6 | 516 | 80 | |
GenBank mRNA | AK036528.1 | - | 5 - 6 | 480 | 80 | |
AK053578.1 | - | 5 - 6 | 478 | 80 | ||
AK089460.1 | - | 5 - 6 | 473 | 80 | ||
AK139284.1 | - | 5 - 6 | 463 | 80 | ||
AK151561.1 | - | 5 - 6 | 480 | 80 | ||
AK152639.1 | - | 5 - 6 | 480 | 80 | ||
AK160584.1 | - | 5 - 6 | 490 | 80 | ||
AK164357.1 | - | 5 - 6 | 479 | 80 | ||
AK170955.1 | - | 5 - 6 | 465 | 80 | ||
AK171198.1 | - | 5 - 6 | 494 | 80 | ||
BC023245.1 | - | 5 - 6 | 469 | 4459337 | 80 | |
BC060709.1 | - | 5 - 6 | 471 | 6409692 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |