Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026159.4 | NP_080435.3 | 2 - 3 | 347 | 77 | |
GenBank mRNA | AK144115.1 | - | 503 | 77 | ||
AY704159.1 | - | 2 - 3 | 347 | 77 | ||
BC117750.1 | - | 2 - 3 | 342 | 77 | ||
BC117751.1 | - | 2 - 3 | 347 | 40090531 | 77 | |
EF620363.1 | - | 373 | 77 | |||
EF620364.1 | - | 373 | 77 | |||
EF620365.1 | - | 2 - 3 | 373 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |