Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145937.3 | NP_666049.2 | 2 - 3 | 468 | 70 | |
GenBank mRNA | AK151874.1 | - | 2 - 3 | 469 | 70 | |
AK159192.1 | - | 2 - 3 | 457 | 70 | ||
AK160917.1 | - | 2 - 3 | 450 | 70 | ||
AK161203.1 | - | 2 - 3 | 461 | 70 | ||
AK194677.1 | - | 1 - 2 | 47 | 70 | ||
AK195132.1 | - | 1 - 2 | 47 | 70 | ||
AK198586.1 | - | 1 - 2 | 70 | |||
BC026981.1 | - | 2 - 3 | 441 | 5365580 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |