Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 18 - 19 | 3057 | 71 | |
GenBank mRNA | AK052087.1 | - | 2 - 3 | 1038 | 71 | |
AK135085.1 | - | 3 - 4 | 385 | 71 | ||
AK144877.1 | - | 18 - 19 | 3028 | 71 | ||
AK166055.1 | - | 18 - 19 | 3038 | 71 | ||
BC011506.3 | - | 18 - 19 | 2991 | 71 | ||
BC022581.1 | - | 3 - 4 | 332 | 71 | ||
BC080691.1 | - | 18 - 19 | 2743 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |