Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175526.3 | NP_780735.2 | 1 - 2 | 392 | 91 | |
GenBank mRNA | AK031121.1 | - | 1 - 2 | 214 | 91 | |
AK036846.1 | - | 1 - 2 | 247 | 91 | ||
AK049608.1 | - | 1 - 2 | 139 | 91 | ||
AK165920.1 | - | 1 - 2 | 251 | 91 | ||
BC052840.1 | - | 1 - 2 | 251 | 30050488 | 91 | |
BX512526.1 | - | 1 - 2 | 392 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |