Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164079.1 | NP_001157551.1 | 2 - 3 | 399 | 83 | |
NM_011585.4 | NP_035715.1 | 2 - 3 | 399 | 83 | ||
GenBank mRNA | AK009502.1 | - | 1 - 2 | 72 | 83 | |
AK009914.1 | - | 1 - 2 | 72 | 83 | ||
AK028340.1 | - | 1 - 2 | 72 | 83 | ||
AK088486.1 | - | 2 - 3 | 335 | 83 | ||
AK144222.1 | - | 2 - 3 | 352 | 83 | ||
BC023813.1 | - | 2 - 3 | 314 | 5353938 | 83 | |
BC027826.1 | - | 2 - 3 | 241 | 83 | ||
U00689.1 | - | 2 - 3 | 340 | 83 | ||
U55862.1 | - | 2 - 3 | 207 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |