Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024251.4 | NP_077213.2 | 1 - 2 | 183 | 110 | |
GenBank mRNA | AK034191.1 | - | 1 - 2 | 163 | 110 | |
BC002179.1 | - | 1 - 2 | 103 | 3487226 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |