Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600007823.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007823.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007823.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007823.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007823.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007832.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007832.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007832.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007832.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007832.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007827.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007827.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007827.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007827.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007827.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007829.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007829.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007829.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007829.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007829.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007822.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007822.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007822.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007822.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007822.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007820.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007820.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007820.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007820.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007820.mm.1 |
NM_020045.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007831.mm.1 |
TC0600001001.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007831.mm.1 |
NM_001170591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007831.mm.1 |
NM_001170591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007831.mm.1 |
NM_020045.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007831.mm.1 |
NM_020045.3 |
Refseq |