Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197985.3 | NP_932102.2 | 3 - 4 | 442 | 79 | |
GenBank mRNA | AK039667.1 | - | 3 - 4 | 650 | 79 | |
AK134787.1 | - | 3 - 4 | 458 | 79 | ||
AK148295.1 | - | 3 - 4 | 584 | 79 | ||
AK160534.1 | - | 3 - 4 | 459 | 79 | ||
AY424291.1 | - | 3 - 4 | 442 | 79 | ||
BC024094.1 | - | 3 - 4 | 442 | 5132696 | 79 | |
BC064109.1 | - | 3 - 4 | 440 | 3371697 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |