Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026267.2 | NP_080543.2 | 1 - 2 | 108 | 59 | |
GenBank mRNA | AK004805.1 | - | 1 - 2 | 109 | 59 | |
AK005263.1 | - | 1 - 2 | 99 | 59 | ||
AK081598.1 | - | 1 - 2 | 109 | 59 | ||
AK168500.1 | - | 1 - 2 | 109 | 59 | ||
AK220236.1 | - | 1 - 2 | 94 | 59 | ||
BC011466.1 | - | 1 - 2 | 107 | 4013043 | 59 | |
BC119133.1 | - | 1 - 2 | 120 | 8733882 | 59 | |
BC120841.1 | - | 1 - 2 | 191 | 40129764 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |