Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168256.1 | NP_001161728.1 | 9 - 10 | 721 | 85 | |
NM_001168257.1 | NP_001161729.1 | 8 - 9 | 646 | 85 | ||
NM_001168258.1 | NP_001161730.1 | 4 - 5 | 446 | 85 | ||
NM_001168259.1 | NP_001161731.1 | 5 - 6 | 213 | 85 | ||
NM_144805.2 | NP_659054.2 | 9 - 10 | 589 | 85 | ||
GenBank mRNA | AK078864.1 | - | 5 - 6 | 214 | 85 | |
BX511925.1 | - | 4 - 5 | 173 | 85 | ||
CT010303.1 | - | 8 - 9 | 519 | 85 | ||
CV561368.1 | - | 8 - 9 | 588 | 85 | ||
CX564997.1 | - | 9 - 10 | 620 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |