Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027526.1 | NP_081802.1 | 2 - 3 | 284 | 83 | |
GenBank mRNA | AK018120.1 | - | 1 - 2 | 115 | 83 | |
AK158756.1 | - | 3 - 4 | 310 | 83 | ||
BC138284.1 | - | 2 - 3 | 250 | 8861303 | 83 | |
BC145232.1 | - | 2 - 3 | 250 | 9053784 | 83 | |
BC145752.1 | - | 2 - 3 | 250 | 40131055 | 83 | |
BY256830.1 | - | 2 - 3 | 232 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |