Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 18 - 19 | 3187 | 118 | |
GenBank mRNA | AK129082.1 | - | 18 - 19 | 3147 | 118 | |
AK133013.1 | - | 18 - 19 | 3137 | 118 | ||
AK162178.1 | - | 18 - 19 | 3245 | 118 | ||
BC030906.1 | - | 11 - 12 | 1987 | 118 | ||
BC057054.1 | - | 18 - 19 | 3187 | 6827225 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |