Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 21 - 22 | 3631 | 94 | |
GenBank mRNA | AK129082.1 | - | 21 - 22 | 3591 | 94 | |
AK133013.1 | - | 21 - 22 | 3581 | 94 | ||
AK162178.1 | - | 21 - 22 | 3691 | 94 | ||
BC030906.1 | - | 14 - 15 | 2431 | 94 | ||
BC057054.1 | - | 21 - 22 | 3631 | 6827225 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |