Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194339.1 | NP_919320.1 | 7 - 8 | 1067 | 108 | |
GenBank mRNA | AK028266.1 | - | 7 - 8 | 1066 | 108 | |
AK129082.1 | - | 7 - 8 | 1027 | 108 | ||
AK133013.1 | - | 7 - 8 | 1017 | 108 | ||
AK134315.1 | - | 7 - 8 | 1075 | 108 | ||
AK135031.1 | - | 6 - 7 | 1564 | 108 | ||
AK162178.1 | - | 7 - 8 | 1125 | 108 | ||
BC057054.1 | - | 7 - 8 | 1067 | 6827225 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |