Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164195.1 | NP_001157667.1 | 3 - 4 | 532 | 83 | |
NM_019961.3 | NP_064345.1 | 3 - 4 | 532 | 83 | ||
GenBank mRNA | AF152996.1 | - | 3 - 4 | 455 | 83 | |
AK005315.1 | - | 3 - 4 | 388 | 83 | ||
AK005978.1 | - | 2 - 3 | 229 | 83 | ||
AK012827.1 | - | 3 - 4 | 511 | 83 | ||
AK146934.1 | - | 3 - 4 | 511 | 83 | ||
BC033415.1 | - | 3 - 4 | 414 | 5008068 | 83 | |
BC037606.1 | - | 3 - 4 | 495 | 5361347 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |