Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164195.1 | NP_001157667.1 | 8 - 9 | 984 | 104 | |
NM_019961.3 | NP_064345.1 | 8 - 9 | 984 | 104 | ||
GenBank mRNA | AF152996.1 | - | 8 - 9 | 907 | 104 | |
AK005315.1 | - | 8 - 9 | 840 | 104 | ||
AK005978.1 | - | 7 - 8 | 681 | 104 | ||
AK012827.1 | - | 8 - 9 | 103 | |||
AK075820.1 | - | 8 - 9 | 899 | 104 | ||
AK083778.1 | - | 3 - 4 | 471 | 104 | ||
AK146934.1 | - | 8 - 9 | 963 | 104 | ||
BC033415.1 | - | 8 - 9 | 866 | 5008068 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |