Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028273.1 | - | 3 - 4 | 948 | 75 | |
AK031129.1 | - | 3 - 4 | 928 | 75 | ||
AK044709.1 | - | 3 - 4 | 945 | 75 | ||
AK044728.1 | - | 3 - 4 | 938 | 75 | ||
AK163571.1 | - | 3 - 4 | 947 | 75 | ||
AK169554.1 | - | 2 - 3 | 172 | 75 | ||
BC057601.1 | - | 3 - 4 | 387 | 6403474 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |