Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028273.1 | - | 6 - 7 | 1368 | 91 | |
AK044709.1 | - | 6 - 7 | 1535 | 91 | ||
AK053568.1 | - | 4 - 5 | 2755 | 91 | ||
BC057601.1 | - | 7 - 8 | 813 | 6403474 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |