Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031633.1 | - | 4 - 5 | 410 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |