Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026257.3 | NP_080533.1 | 3 - 4 | 197 | 66 | |
NR_045571.1 | 3 - 4 | 197 | 66 | |||
GenBank mRNA | AK015720.1 | - | 3 - 4 | 181 | 66 | |
AK048494.1 | - | 3 - 4 | 198 | 66 | ||
BC019461.1 | - | 3 - 4 | 174 | 4224033 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |