Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134228.1 | NP_598989.1 | 1 | 340 | 74 | |
GenBank mRNA | AF394955.1 | - | 1 | 340 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |