Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009185.3 | NP_033211.2 | 3 - 4 | 420 | 74 | |
GenBank mRNA | AK049223.1 | - | 3 - 4 | 357 | 74 | |
AK049550.1 | - | 3 - 4 | 357 | 74 | ||
BC049865.1 | - | 3 - 4 | 374 | 74 | ||
BC060706.1 | - | 3 - 4 | 335 | 74 | ||
BC087545.1 | - | 3 - 4 | 359 | 74 | ||
BC108373.1 | - | 3 - 4 | 335 | 74 | ||
BC141175.1 | - | 3 - 4 | 293 | 9055787 | 74 | |
BC145493.1 | - | 3 - 4 | 293 | 9054056 | 74 | |
CJ098395.1 | - | 3 - 4 | 421 | 74 | ||
CJ098467.1 | - | 3 - 4 | 421 | 74 | ||
U36778.1 | - | 3 - 4 | 338 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |