Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1436040_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436040_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436040_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0400012419.mm.1 |
NR_029468.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012419.mm.1 |
NR_029468.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012419.mm.1 |
TC0400001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400012432.mm.1 |
NR_029468.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012432.mm.1 |
NR_029468.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012432.mm.1 |
TC0400001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400012425.mm.1 |
NR_029468.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012425.mm.1 |
NR_029468.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012425.mm.1 |
TC0400001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400012418.mm.1 |
NR_029468.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012418.mm.1 |
NR_029468.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012418.mm.1 |
TC0400001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400012433.mm.1 |
NR_029468.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012433.mm.1 |
NR_029468.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400012433.mm.1 |
TC0400001582.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1436040_at |
- |
- |
Mouse Expression Set 430 |
1436040_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1436040_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1436040_PM_at |
- |
- |