Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034030.2 | NP_001029202.1 | 1 - 2 | 90 | 67 | |
GenBank mRNA | AB012092.1 | - | 1 - 2 | 90 | 67 | |
AB012291.1 | - | 1 - 2 | 90 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |