Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190408.1 | NP_001177337.1 | 4 - 5 | 971 | 113 | |
NM_030228.3 | NP_084504.2 | 5 - 6 | 1229 | 113 | ||
NM_144560.3 | NP_653146.1 | 5 - 6 | 1229 | 113 | ||
GenBank mRNA | AF508323.1 | - | 3 - 4 | 873 | 113 | |
AF508324.1 | - | 3 - 4 | 873 | 113 | ||
AK019661.1 | - | 5 - 6 | 1204 | 113 | ||
AK052229.1 | - | 5 - 6 | 1230 | 113 | ||
BC031785.1 | - | 4 - 5 | 921 | 4194333 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |